Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ACSL4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587082
Description
Purity is > 95% by SDS-PAGE.
The protein encoded by this gene is an isozyme of the long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation. This isozyme preferentially utilizes arachidonate as substrate. The absence of this enzyme may contribute to the mental retardation or Alport syndrome. Alternative splicing of this gene generates 2 transcript variants.
Specifications
ACSL4 | |
Polyclonal | |
Unconjugated | |
Acsl4 | |
9430020A05Rik; Acs4; ACSL4; acyl-CoA synthetase 4; acyl-CoA synthetase long-chain family member 4; Arachidonate--CoA ligase; AU018108; Facl4; fatty acid Coenzyme A ligase, long chain 4; fatty acid-Coenzyme A ligase long chain 4; fatty acid-Coenzyme A ligase, long chain 4; fatty-acid-Coenzyme A ligase, long-chain 4; L MRX63; LACS 4; Lacs4; lignoceroyl-CoA synthase; long-chain acyl-CoA synthetase 4; long-chain fatty-acid-Coenzyme A ligase 4; long-chain-fatty-acid--CoA ligase 4; mACS4; MRX63; MRX68 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2182, 50790 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O60488, Q9QUJ7 | |
Acsl4 | |
Recombinant protein of human ACSL4. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction