Learn More
Invitrogen™ ACSL4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA593240
Description
Immunogen sequence: MAKRIKAKPT SDKPGSPYRS VTHFDSLAVI DIPGADTLDK LFDHAVSKFG KKDSLGTREI LSEENEMQPN GKVFKKLILG NYKWMNYLEV NRRVNNFGSG LTALGLKPKN TIAIFCETRA EWMIAAQTCF KYNFPLVTLY ATLGKEAVVH GLNESEASYL ITSVELLESK LKTALLDISC VKHIIYVDNK AINKAEYPEG FEIHSMQSVE ELGSNPENLG IPPSRPTPSD MAIVMYTSGS TGRPKGVMMH HSNLIAGMTG QCERIPGLGP KDTYIGYLPL; Positive Samples: HeLa, HT-1080, 22Rv1, Mouse liver; Cellular Location: Endoplasmic reticulum membrane, Microsome membrane, Mitochondrion outer membrane, Peroxisome membrane, Single-pass type III membrane protein.
The protein encoded by this gene is an isozyme of the long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation. This isozyme preferentially utilizes arachidonate as substrate. The absence of this enzyme may contribute to the mental retardation or Alport syndrome. Alternative splicing of this gene generates 2 transcript variants.
Specifications
ACSL4 | |
Polyclonal | |
Unconjugated | |
Acsl4 | |
9430020A05Rik; Acs4; ACSL4; acyl-CoA synthetase 4; acyl-CoA synthetase long-chain family member 4; Arachidonate--CoA ligase; AU018108; Facl4; fatty acid Coenzyme A ligase, long chain 4; fatty acid-Coenzyme A ligase long chain 4; fatty acid-Coenzyme A ligase, long chain 4; fatty-acid-Coenzyme A ligase, long-chain 4; L MRX63; LACS 4; Lacs4; lignoceroyl-CoA synthase; long-chain acyl-CoA synthetase 4; long-chain fatty-acid-Coenzyme A ligase 4; long-chain-fatty-acid--CoA ligase 4; mACS4; MRX63; MRX68 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
113976, 2182, 50790 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
1.26 mg/mL | |
PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
O35547, O60488, Q9QUJ7 | |
Acsl4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 42-321 of human ACSL4 (NP_0752661?). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.