Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACSL5 Monoclonal Antibody (CL0275)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA524595

Catalog No. PIMA524595


Only null left
Add to Cart

Description

Description

Immunogen sequence: VHIVNKADIA MVICDTPQKA LVLIGNVEKG FTPSLKVIIL MDPFDDDLKQ RGEKSGIEIL SLYDAENLGK EHFRKPVPPS PEDLSVICFT SGTTGDPK Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 55%. Binds to an epitope located within the peptide sequence MDPFDDDLKQ as determined by overlapping synthetic peptides.

The protein encoded by this gene is an isozyme of the long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation. This isozyme is highly expressed in uterus and spleen, and in trace amounts in normal brain, but has markedly increased levels in malignant gliomas. This gene functions in mediating fatty acid-induced glioma cell growth. Three transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ACSL5
Monoclonal
1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9ULC5
ACSL5
Recombinant protein corresponding to Human ACSL5.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
CL0275
Unconjugated
ACSL5
1700030F05Rik; ACS2; ACS5; Acsl5; acyl-CoA synthetase 5; acyl-CoA synthetase long chain family member 5; acyl-CoA synthetase long-chain family member 5; Arachidonate--CoA ligase; FACL5; FACL5 for fatty acid coenzyme A ligase 5; fatty acid coenzyme A ligase 5; fatty acid Coenzyme A ligase, long chain 5; fatty-acid-Coenzyme A ligase, long-chain 5; HGNC:16526; LACS 5; Long-chain acyl-CoA synthetase 5; long-chain fatty acid coenzyme A ligase 5; long-chain-fatty-acid--CoA ligase 5; UNQ633/PRO1250; zgc:92083
Mouse
Protein A
RUO
51703
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.