Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACSL6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599052
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599052 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599052 Supplier Invitrogen™ Supplier No. PA599052
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ACSL6 Polyclonal Antibody for Western Blot, IHC (P), ELISA

FACL6 is involved in activation of long-chain fatty acids for both synthesis of cellular lipids, and degradation via beta-oxidation. It plays an important role in fatty acid metabolism in brain and the acyl-CoAs produced may be utilized exclusively for the synthesis of the brain lipid. FACL6 is expressed predominantly in erythrocyte precursors, in particular in reticulocytes, fetal blood cells derieved from fetal liver, haemopoietic stem cells from cord blood, bone marrow, and brain. Expression is low at earlier stages of erythroid development but is very high in reticulocytes. This protein is involved in myelodysplastic syndrome (MDS) with basophilia, acute myelogenous leukemia (AML) with eosinophilia, and acute eosinophilic leukemia (AEL). It is characterized by a chromosomal translocation t(5;12)(q31;p13) that involves ETV6 and ACSL6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACSL6
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.46 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Acsl6
Gene Accession No. Q91WC3, Q9UKU0
Gene Alias A330035H04Rik; ACS2; ACSL6; acyl-CoA synthetase long-chain family member 6; Arachidonate--CoA ligase; AW050338; FACL6; fatty acid Coenzyme A ligase, long chain 6; fatty-acid-Coenzyme A ligase, long-chain 6; KIAA0837; L LACS5; LACS; LACS 6; LACS2; LACS5; Lacsl; long fatty acyl-CoA synthetase 2; long-chain acyl-CoA synthetase 6; Long-chain-fatty-acid--CoA ligase 6; long-chain-fatty-acid--CoA ligase, brain isozyme; mKIAA0837
Gene Symbols Acsl6
Host Species Rabbit
Immunogen Recombinant Human Long-chain-fatty-acid--CoA ligase 6 protein (458-697AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216739, 23305
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.