Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACSM1 Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Catalog No. FEPA5145453
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145453 200 μL
FEPA5145454 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEPA5145453 Supplier Invitrogen™ Supplier No. PA5145453
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ACSM1 has medium-chain fatty acid:CoA ligase activity with broad substrate specificity (in vitro). This protein acts on acids from C(4) to C(11) and on the corresponding 3-hydroxy-and 2,3-or 3,4-unsaturated acids (in vitro). This protein functions as GTP-dependent lipoate-activating enzyme that generates the substrate for lipoyltransferase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACSM1
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene ACSM1
Gene Accession No. Q08AH1, Q91VA0
Gene Alias ACSM1; Acyl-CoA synthetase medium-chain family member 1; acyl-coenzyme A synthetase ACSM1, mitochondrial; Benzoate--CoA ligase; BUCS1; Butyrate CoA ligase; butyrate--CoA ligase 1; butyryl Coenzyme A synthetase 1; butyryl-coenzyme A synthetase 1; LAE; Lipoate-activating enzyme; MACS1; medium-chain acyl-CoA synthetase; middle-chain acyl-CoA synthetase 1; Xenobiotic/medium-chain fatty acid-CoA ligase HXM-B
Gene Symbols ACSM1
Host Species Rabbit
Immunogen Recombinant Human ACSM1 protein, Ser370-Pro557 (Accession #Q08AH1).
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116285, 117147, 361638
Target Species Human, Mouse, Pig, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.