Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACSM2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570859
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570859 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570859 Supplier Invitrogen™ Supplier No. PA570859
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 85%; Dog: 93%; Guinea Pig: 86%; Horse: 85%; Human: 100%; Mouse: 86%; Rabbit: 93%; Rat: 90%; Zebrafish: 85%.

This gene encodes a mitochondrial acyl-coenzyme A synthetase that is specific for medium chain fatty acids. These enzymes catalyze fatty acid activation, the first step of fatty acid metabolism, through the transfer of acyl-CoA. These enzymes also participate in the glycine conjugation pathway in the detoxification of xenobiotics such as benzoate and ibuprofen. Expression levels of this gene in the kidney may be correlated with kidney function. This gene and its paralog ACSM2B (Gene ID: 348158), both present on chromosome 16, likely arose from a chromosomal duplication event.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACSM2A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ACSM2A
Gene Accession No. Q08AH3
Gene Alias A-923A4.1; ACSM2; ACSM2A; acyl-CoA synthetase medium-chain family member 2; acyl-CoA synthetase medium-chain family member 2A; Acyl-coenzyme A synthetase ACSM2A, mitochondrial; Benzoate--CoA ligase; Butyrate--CoA ligase 2A; butyryl-coenzyme A synthetase 2A; Homolog of rat kidney-specific (KS); MACS2; Middle-chain acyl-CoA synthetase 2A
Gene Symbols ACSM2A
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human ACSM2A.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 123876
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.