Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ACSM2A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590559
Description
Immunogen sequence: RPIGIFSGYV DNPDKTAANI RGDFWLLGDR GIKDEDGYFQ FMGRANDIIN SSGYRIGPSE VENALMEHPA VVETAVISSP DPVRGEVVKA FVVLASQFLS HDPEQLTKEL QQHVKSVTAP YKYPRKIEFV LNLPKTVTGK IQRAKLRDKE WKMSGKARAQ.
This gene encodes a mitochondrial acyl-coenzyme A synthetase that is specific for medium chain fatty acids. These enzymes catalyze fatty acid activation, the first step of fatty acid metabolism, through the transfer of acyl-CoA. These enzymes also participate in the glycine conjugation pathway in the detoxification of xenobiotics such as benzoate and ibuprofen. Expression levels of this gene in the kidney may be correlated with kidney function. This gene and its paralog ACSM2B (Gene ID: 348158), both present on chromosome 16, likely arose from a chromosomal duplication event.
Specifications
ACSM2A | |
Polyclonal | |
Unconjugated | |
A-923A4.1; ACSM2; ACSM2A; acyl-CoA synthetase medium-chain family member 2; acyl-CoA synthetase medium-chain family member 2A; Acyl-coenzyme A synthetase ACSM2A, mitochondrial; Benzoate--CoA ligase; Butyrate--CoA ligase 2A; butyryl-coenzyme A synthetase 2A; Homolog of rat kidney-specific (KS); MACS2; Middle-chain acyl-CoA synthetase 2A | |
Recombinant fusion protein containing a sequence corresponding to amino acids 418-577 of human ACSM2A (NP_0012951011). | |
100 μL | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.61 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction