Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACSM2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571436
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571436 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571436 Supplier Invitrogen™ Supplier No. PA571436
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ACSM2B Polyclonal Antibody for Western Blot

ACSM2B gene ontology annotations related to this gene include catalytic activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACSM2B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ACSM2B
Gene Accession No. Q68CK6
Gene Alias ACSM2; ACSM2B; acyl-CoA synthetase medium-chain family member 2; acyl-CoA synthetase medium-chain family member 2B; Acyl-coenzyme A synthetase ACSM2B, mitochondrial; Benzoate--CoA ligase; Butyrate--CoA ligase 2B; butyryl-coenzyme A synthetase 2B; HXMA; HYST1046; Middle-chain acyl-CoA synthetase 2B; xenobiotic/medium-chain fatty acid:CoA ligase; xenobiotic/medium-chain fatty acid-CoA ligase HXM-A
Gene Symbols ACSM2B
Host Species Rabbit
Immunogen synthetic peptide directed towards the C terminal region of human ACSM2B.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 348158
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.