Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACSM3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | ACSM3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ACSM3 Polyclonal specifically detects ACSM3 in Human samples. It is validated for Western Blot.Specifications
ACSM3 | |
Polyclonal | |
Rabbit | |
Q53FZ2 | |
6296 | |
Synthetic peptides corresponding to thetase medium chain family member 3 Antibody against the N terminal of ACSM3. Immunizing peptide sequence SMKQDFKLGIPEYFNFAKDVLDQWTDKEKAGKKPSNPAFWWINRNGEEMR. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
acyl-CoA synthetase medium-chain family member 3Protein SA homolog, acyl-coenzyme A synthetase ACSM3, mitochondrial, Butyrate--CoA ligase 3, Butyryl-coenzyme A synthetase 3, EC 6.2.1, Middle-chain acyl-CoA synthetase 3, SA, SA (rat hypertension-associated) homolog, SA hypertension-associated homolog, SA hypertension-associated homolog (rat), SAH, SAHEC 6.2.1.2 | |
ACSM3 | |
IgG | |
66 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title