Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACSM3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112973
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112973 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112973 Supplier Invitrogen™ Supplier No. PA5112973
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ACSM3 Polyclonal Antibody for ELISA, IHC (P), Western Blot

Has medium-chain fatty acid:CoA ligase activity with broad substrate specificity (in vitro). Acts on acids from C(4) to C(11) and on the corresponding 3-hydroxy- and 2,3- or 3,4-unsaturated acids (in vitro).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACSM3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.461 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ACSM3
Gene Accession No. Q3UNX5, Q53FZ2
Gene Alias Acsm3; acyl-CoA synthetase medium chain family member 3; acyl-CoA synthetase medium-chain family member 3; Acyl-coenzyme A synthetase ACSM3, mitochondrial; butyrate--CoA ligase 3; butyryl coenzyme A synthetase 3; butyryl-coenzyme A synthetase 3; CH73-60L17.3; Middle-chain acyl-CoA synthetase 3; Propionate--CoA ligase; Protein SA; protein SA homolog; Sa; SA (rat hypertension-associated) homolog; SA hypertension-associated homolog; SA rat hypertension-associated homolog; SA rat hypertension-associated protein; SAH; ssubA; zgc:172040
Gene Symbols ACSM3
Host Species Rabbit
Immunogen Recombinant Human Acyl-coenzyme A synthetase ACSM3, mitochondrial protein (387-586AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20216, 6296
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.