Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACSM3 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS7641R

Description
Has medium-chain fatty acid:CoA ligase activity with broad substrate specificity (in vitro). Acts on acids from C(4) to C(11) and on the corresponding 3-hydroxy- and 2,3- or 3,4-unsaturated acids (in vitro).Specifications
ACSM3 | |
Polyclonal | |
Unconjugated | |
ACSM3 | |
ACSM3; acyl-CoA synthetase medium chain family member 3; Acyl-CoA synthetase medium-chain family member 3; Acyl-coenzyme A synthetase ACSM3, mitochondrial; Butyrate--CoA ligase 3; butyryl coenzyme A synthetase 3; Butyryl-coenzyme A synthetase 3; CH73-60L17.3; LOC100345414; Middle-chain acyl-CoA synthetase 3; Propionate--CoA ligase; Protein SA; Protein SA homolog; Sa; SA (rat hypertension-associated) homolog; SA hypertension-associated homolog; SA rat hypertension-associated homolog; SA rat hypertension-associated protein; SAH; ssubA; zgc:172040 | |
Rabbit | |
Protein A | |
RUO | |
6296 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q53FZ2 | |
ACSM3 | |
KLH conjugated synthetic peptide derived from human ACSM3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction