Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACSM4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562082
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562082 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562082 Supplier Invitrogen™ Supplier No. PA562082
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FRYQTFRFIW LTKPPGRRLH KDHQLWTPLT LADFEAINRC NRPLPKNFNF AADV Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 74%.

Has medium-chain fatty acid:CoA ligase activity with broad substrate specificity (in vitro). Acts on acids from C(4) to C(11) and on the corresponding 3-hydroxy- and 2,3- or 3,4- unsaturated acids (in vitro).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACSM4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ACSM4
Gene Accession No. P0C7M7
Gene Alias ACSM4; acyl-CoA synthetase medium chain family member 4; acyl-CoA synthetase medium-chain family member 4; Acyl-coenzyme A synthetase ACSM4, mitochondrial; Olfactory specific medium-chain acyl CoA synthetase; Omacs; O-MACS
Gene Symbols ACSM4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ACSM4. Recombinant protein control fragment (Product #RP-100781).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 341392
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.