Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ACSM4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570980
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
Has medium-chain fatty acid:CoA ligase activity with broad substrate specificity (in vitro). Acts on acids from C(4) to C(11) and on the corresponding 3-hydroxy- and 2,3- or 3,4- unsaturated acids (in vitro).
Specifications
ACSM4 | |
Polyclonal | |
Unconjugated | |
ACSM4 | |
ACSM4; acyl-CoA synthetase medium chain family member 4; acyl-CoA synthetase medium-chain family member 4; Acyl-coenzyme A synthetase ACSM4, mitochondrial; Olfactory specific medium-chain acyl CoA synthetase; Omacs; O-MACS | |
Rabbit | |
Affinity chromatography | |
RUO | |
341392 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P0C7M7 | |
ACSM4 | |
synthetic peptide directed towards the middle region of human ACSM4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction