Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACSM5 Monoclonal Antibody (OTI1G1), TrueMAB™, OriGene
SDP

Catalog No. 502540323
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0323 30 μL
50-167-5133 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0323 Supplier OriGene Supplier No. TA504441S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ACSM5 Monoclonal antibody specifically detects ACSM5 in Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen ACSM5
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI1G1
Concentration 0.81 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Schip1
Gene Accession No. P0DPB3
Gene Alias Aa2-174; ACSM5; Acyl-CoA synthetase medium-chain family member 5; acyl-coenzyme A synthetase ACSM5, mitochondrial; C730019D22; C730027J19Rik; Cc1-38; MACS3; RGD1309578
Host Species Mouse
Immunogen Full length human recombinant protein of human SCHIP1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29970
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.