Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACSM5 Monoclonal Antibody (OTI4B11)
GREENER_CHOICE

Catalog No. PIMA525918
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525918 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525918 Supplier Invitrogen™ Supplier No. MA525918
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ACSM5 Monoclonal Antibody for Western Blot, Flow

Catalyzes the activation of fatty acids by CoA to produce an acyl-CoA, the first step in fatty acid metabolism. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACSM5
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI4B11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ACSM5
Gene Accession No. Q6NUN0
Gene Alias Aa2-174; ACSM5; Acyl-CoA synthetase medium-chain family member 5; acyl-coenzyme A synthetase ACSM5, mitochondrial; C730019D22; C730027J19Rik; Cc1-38; MACS3; RGD1309578
Gene Symbols ACSM5
Host Species Mouse
Immunogen Full length human recombinant protein of ACSM5 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54988
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.