Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACSM5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550299
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550299 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550299 Supplier Invitrogen™ Supplier No. PA550299
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total ACSM5 protein.

Catalyzes the activation of fatty acids by CoA to produce an acyl-CoA, the first step in fatty acid metabolism. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACSM5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene ACSM5
Gene Accession No. Q6NUN0
Gene Alias Aa2-174; ACSM5; Acyl-CoA synthetase medium-chain family member 5; acyl-coenzyme A synthetase ACSM5, mitochondrial; C730019D22; C730027J19Rik; Cc1-38; MACS3; RGD1309578
Gene Symbols ACSM5
Host Species Rabbit
Immunogen Full length fusion protein of Human ACSM5.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54988
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.