Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACSS1 Monoclonal Antibody (4E12F1)
GREENER_CHOICE

Catalog No. FERMA548794
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548794 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548794 Supplier Invitrogen™ Supplier No. MA548794
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ACSS1 Monoclonal Antibody for Western Blot, Flow, ELISA

This gene encodes a mitochondrial acetyl-CoA synthetase enzyme. A similar protein in mice plays an important role in the tricarboxylic acid cycle by catalyzing the conversion of acetate to acetyl CoA. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACSS1
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 4E12F1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene ACSS1
Gene Accession No. Q9NUB1
Gene Alias 1110032O15Rik; Acas2; ACAS2L; ACECS1; AceCS2; AceCS2L; acetate--CoA ligase 2; Acetyl-CoA synthetase 2; acetyl-coenzyme A synthetase 2-like, mitochondrial; ACSS1; Acyl-CoA synthetase short-chain family member 1; AI788978; KIAA1846; Propionate--CoA ligase
Gene Symbols ACSS1
Host Species Mouse
Immunogen Purified recombinant fragment of human ACSS1 (AA: 548-689) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84532
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.