Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACSS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101399
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101399 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101399 Supplier Invitrogen™ Supplier No. PA5101399
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ACSS1.

This gene encodes a mitochondrial acetyl-CoA synthetase enzyme. A similar protein in mice plays an important role in the tricarboxylic acid cycle by catalyzing the conversion of acetate to acetyl CoA. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACSS1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ACSS1
Gene Accession No. Q99NB1, Q9NUB1
Gene Alias 1110032O15Rik; Acas2; ACAS2L; ACECS1; AceCS2; AceCS2L; acetate--CoA ligase 2; Acetyl-CoA synthetase 2; acetyl-coenzyme A synthetase 2-like, mitochondrial; ACSS1; Acyl-CoA synthetase short-chain family member 1; AI788978; KIAA1846; Propionate--CoA ligase
Gene Symbols ACSS1
Host Species Rabbit
Immunogen A synthesized peptide derived from human ACSS1(Accession Q9NUB1), corresponding to amino acid residues T618-P668.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 68738, 84532
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.