Learn More
Invitrogen™ ACSS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109357
Description
Immunogen sequence: ITGHSYVVYG PLCNGATSVL FESTPVYPNA GRYWETVERL KINQFYGAPT AVRLLLKYGD AWVKKYDRSS LRTLGSVGEP INCEAWEWLH RVVGDSRCTL VDTWWQTETG GICIAPRPSE EGAEILPAMA MRPFFGIVPV LMDEKGSVVE GSNVSGALCI SQAWPGMART IYGDHQRFVD AYFKAYPGYY FTGDGAYRTE GGYYQITGRM DDVINISGHR LGTAEIEDAI ADHPAVPESA VIGYPHDIKG EAAFAFIVVK DSAGDSDVVV QELKSMVATK IAKYAVPDEI LVVKRLPKTR SGKVMRRLLR KIITSEAQEL GDTTTLEDPS IIAEILSVYQ KCKDKQAAAK.
This gene encodes a mitochondrial acetyl-CoA synthetase enzyme. A similar protein in mice plays an important role in the tricarboxylic acid cycle by catalyzing the conversion of acetate to acetyl CoA. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
ACSS1 | |
Polyclonal | |
Unconjugated | |
ACSS1 | |
1110032O15Rik; Acas2; ACAS2L; ACECS1; AceCS2; AceCS2L; acetate--CoA ligase 2; Acetyl-CoA synthetase 2; acetyl-coenzyme A synthetase 2-like, mitochondrial; ACSS1; Acyl-CoA synthetase short-chain family member 1; AI788978; KIAA1846; Propionate--CoA ligase | |
Rabbit | |
Affinity Chromatography | |
RUO | |
296259, 68738, 84532 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
2.21 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q99NB1, Q9NUB1 | |
ACSS1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 340-689 of human ACSS1 (NP_115890.2). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.