Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACSS2 Monoclonal Antibody (OTI3A8), TrueMAB™, OriGene
SDP

Catalog No. 502540806
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0806 30 μL
50-167-4461 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0806 Supplier OriGene Supplier No. TA503606S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ACSS2 Monoclonal antibody specifically detects ACSS2 in Canine, Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen ACSS2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3A8
Concentration 0.91 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Gtf2f1
Gene Accession No. P35269
Gene Alias 1110017C11Rik; ACAS; Acas1; Acas2; ACECS; AceCS1; acetate thiokinase; acetate-CoA ligase; acetate--CoA ligase; Acetyl-CoA synthetase; Acetyl-CoA synthetase 1; acetyl-Coenzyme A synthetase 1 (AMP forming); acetyl-Coenzyme A synthetase 2 (ADP forming); acetyl-Coenzyme A synthetase 2 (AMP forming); acetyl-coenzyme A synthetase, cytoplasmic; ACS; Acs1; ACSA; ACSS2; Acyl-activating enzyme; acyl-CoA synthetase short-chain family member 2; cytoplasmic acetyl-coenzyme A synthetase; dJ1161H23.1; Propionate--CoA ligase
Host Species Mouse
Immunogen Full length human recombinant protein of human GTF2F1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2962, 476731
Target Species Canine, Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.