Learn More
ACSS2 Mouse anti-Human, Clone: OTI3C6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503632

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a cytosolic enzyme that catalyzes the activation of acetate for use in lipid synthesis and energy generation. The protein acts as a monomer and produces acetyl-CoA from acetate in a reaction that requires ATP. Expression of this gene is regulated by sterol regulatory element-binding proteins, transcription factors that activate genes required for the synthesis of cholesterol and unsaturated fatty acids. Alternative splicing results in multiple transcript variants.Specifications
ACSS2 | |
Monoclonal | |
Unconjugated | |
ACSS2 | |
ACAS2, ACECS, ACS, ACSA, dJ1161H23.1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
55902 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI3C6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9NR19 | |
ACSS2 | |
Full length human recombit protein of human ACSS2 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.