Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACSS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552059
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552059 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552059 Supplier Invitrogen™ Supplier No. PA552059
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PGETTQITYH QLLVQVCQFS NVLRKQGIQK GDRVAIYMPM IPELVVAMLA CARIGALHSI VFAGFSSESL CERILDSSCS LLITTDAFYR GEKLVNLKEL ADEALQKCQE KGFPVRCCIV VKHLGRAELG MGDSTSQSPP IKRSCPDVQ Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 90%.

This gene encodes a protein with a sterol sensing domain (SSD) and seven WD domains. In the presence of cholesterol, this protein binds to sterol regulatory element binding proteins (SREBPs) and mediates their transport from the ER to the Golgi. The SREBPs are then proteolytically cleaved and regulate sterol biosynthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACSS2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Acss2
Gene Accession No. Q9NR19
Gene Alias 1110017C11Rik; ACAS; Acas1; Acas2; ACECS; AceCS1; acetate thiokinase; acetate-CoA ligase; acetate--CoA ligase; Acetyl-CoA synthetase; Acetyl-CoA synthetase 1; acetyl-Coenzyme A synthetase 1 (AMP forming); acetyl-Coenzyme A synthetase 2 (ADP forming); acetyl-Coenzyme A synthetase 2 (AMP forming); acetyl-coenzyme A synthetase, cytoplasmic; ACS; Acs1; ACSA; ACSS2; Acyl-activating enzyme; acyl-CoA synthetase short-chain family member 2; cytoplasmic acetyl-coenzyme A synthetase; dJ1161H23.1; Propionate--CoA ligase
Gene Symbols Acss2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ACSS2. Recombinant protein control fragment (Product #RP-102332).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55902
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.