Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACSS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA526612
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA526612 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA526612 Supplier Invitrogen™ Supplier No. PA526612
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ACSS2 Polyclonal Antibody for Western Blot

This gene encodes a protein with a sterol sensing domain (SSD) and seven WD domains. In the presence of cholesterol, this protein binds to sterol regulatory element binding proteins (SREBPs) and mediates their transport from the ER to the Golgi. The SREBPs are then proteolytically cleaved and regulate sterol biosynthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACSS2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Acss2
Gene Accession No. Q9NR19, Q9QXG4
Gene Alias 1110017C11Rik; ACAS; Acas1; Acas2; ACECS; AceCS1; acetate thiokinase; acetate-CoA ligase; acetate--CoA ligase; Acetyl-CoA synthetase; Acetyl-CoA synthetase 1; acetyl-Coenzyme A synthetase 1 (AMP forming); acetyl-Coenzyme A synthetase 2 (ADP forming); acetyl-Coenzyme A synthetase 2 (AMP forming); acetyl-coenzyme A synthetase, cytoplasmic; ACS; Acs1; ACSA; ACSS2; Acyl-activating enzyme; acyl-CoA synthetase short-chain family member 2; cytoplasmic acetyl-coenzyme A synthetase; dJ1161H23.1; Propionate--CoA ligase
Gene Symbols Acss2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 31-60 amino acids from the N-terminal region of human ACSS2.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55902, 60525
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.