Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACSS3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558750
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558750 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558750 Supplier Invitrogen™ Supplier No. PA558750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EYKTHFAASV TDPERFWGKA AEQISWYKPW TKTLENKHSP STRWFVEGML NICYNAVDRH IENGKGDKIA IIYDSPVTNT KATFTYKEVL EQ Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 89%.

Catalyzes the synthesis of acetyl-CoA from short-chain fatty acids (PubMed:28003429). Propionate is the preferred substrate (PubMed:28003429). Can utilize acetate and butyrate with a much lower affinity. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACSS3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Acss3
Gene Accession No. Q9H6R3
Gene Alias 8430416H19Rik; Acetate--CoA ligase 3; ACSS3; Acyl-CoA synthetase short-chain family member 3; acyl-CoA synthetase short-chain family member 3, mitochondrial; AMP-binding enzyme, 33217; Gm874; Propionate--CoA ligase; RGD1307051
Gene Symbols Acss3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ACSS3. Recombinant protein control fragment (Product #RP-97678).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79611
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.