Learn More
Invitrogen™ ACSS3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116567
Description
Positive Samples: Mouse kidney, Mouse lung, Rat kidney, Rat liver Immunogen sequence: AGAALRALVV PGPRGGLGGR GCRALSSGSG SEYKTHFAAS VTDPERFWGK AAEQISWYKP WTKTLENKHS PSTRWFVEGM LNICYNAVDR HIENGKGDKI AIIYDSPVTN TKATFTYKEV LEQVSKLAGV LVKHGIKKGD TVVIYMPMIP QAMYTMLACA RIGAIHSLIF GGFASKELSS RIDHVKPKVV VTASFGIEPG RRVEYVPLVE EALKIGQHKP DKILIYNRPN MEAVPLAPGR DLDWDEEMAK A.
Catalyzes the synthesis of acetyl-CoA from short-chain fatty acids (PubMed:28003429). Propionate is the preferred substrate (PubMed:28003429). Can utilize acetate and butyrate with a much lower affinity. [UniProt].
Specifications
ACSS3 | |
Polyclonal | |
Unconjugated | |
Acss3 | |
8430416H19Rik; Acetate--CoA ligase 3; ACSS3; Acyl-CoA synthetase short-chain family member 3; acyl-CoA synthetase short-chain family member 3, mitochondrial; AMP-binding enzyme, 33217; Gm874; Propionate--CoA ligase; RGD1307051 | |
Rabbit | |
Affinity chromatography | |
RUO | |
314800, 380660 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.22 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q14DH7 | |
Acss3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 30-280 of human ACSS3 (NP_0788361). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.