Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ACT 462206
GREENER_CHOICESDP

Catalog No. 531950 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-195-0 50 mg
53-191-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 53-195-0 Supplier Tocris Bioscience™ Supplier No. 5319/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent, dual orexin receptor antagonist

ACT 462206 is a potent, dual orexin receptor antagonist (IC50 values are 11 and 60 nM for OX2 and OX1 receptors respectively). Exhibits anxiolytic-like properties in vivo. Brain penetrant.

Specifications

Quantity 50 mg
Product Type ACT 462206
Purity >99%
Molecular Weight (g/mol) 388.48
Inhibitors ACT 462206

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.