Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACTA1 Monoclonal Antibody (3H5)
GREENER_CHOICE

Catalog No. PIMA549215
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549215 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549215 Supplier Invitrogen™ Supplier No. MA549215
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Immunogen sequence is identical to the related mouse and rat sequences. Positive Control - WB: human HepG2 whole cell, human SK-OV-3 whole cell, human PANC-1 whole cell, human Hela whole cell, human A549 whole cell, rat PC-12 whole cell, mouse NIH/3T3 whole cell, mouse HEPA1-6 whole cell. IHC: human bladder cancer tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

The product encoded by this gene belongs to the actin family of proteins, which are highly conserved proteins that play a role in cell motility, structure and integrity. Alpha, beta and gamma actin isoforms have been identified, with alpha actins being a major constituent of the contractile apparatus, while beta and gamma actins are involved in the regulation of cell motility. This actin is an alpha actin that is found in skeletal muscle. Mutations in this gene cause nemaline myopathy type 3, congenital myopathy with excess of thin myofilaments, congenital myopathy with cores, and congenital myopathy with fiber-type disproportion, diseases that lead to muscle fiber defects.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACTA1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 3H5
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene ACTA1
Gene Accession No. P68133, P68134, P68136
Gene Alias AA959943; ACTA; Acta1; Acta-2; actin alpha 1; actin alpha 1, skeletal muscle; actin, alpha 1, skeletal muscle; actin, alpha skeletal muscle; Actin, alpha skeletal muscle, intermediate form; Acts; Actsk-1; alpha actin 1; alpha-actin-1; ASMA; beta actin; CFTD; CFTD1; CFTDM; cytoplasmic beta-actin; LOW QUALITY PROTEIN: actin, alpha skeletal muscle; MPFD; NEM1; NEM2; NEM3; nemaline myopathy type 3; SHPM; skeletal alpha actin; unnamed protein product
Gene Symbols ACTA1
Host Species Mouse
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human Actin (277-308aa HETTYNSIMKCDIDIRKDLYANNVMSGGTTMY).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11459, 29437, 58
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.