Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACTA1 Rabbit anti-Human, Mouse, Rat, Unconjugated, Clone: AAOG-1, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB19524
Description
ACTA1 Monoclonal antibody specifically detects ACTA1 in Human, Mouse, Rat samples. It is validated for ICC, WB-Ce, IF, Flow Cyt.Specifications
ACTA1 | |
Monoclonal | |
Unconjugated | |
ACTA1 | |
Rabbit | |
100 μg | |
58 | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
IgG |
Immunocytochemistry, Western Blot, Immunofluorescence, Flow Cytometry | |
AAOG-1 | |
actin, alpha 1, skeletal muscle | |
ACTA, ASMA, CFTD, CFTD1, CFTDM, MPFD, NEM1, NEM2, NEM3 | |
A synthetic peptide corresponding to human ACTA1 | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction