Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ACTA1 Recombinant Rabbit Monoclonal Antibody (4C1)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA550196
Description
ACTA1 Recombinant Monoclonal Antibody for IHC (P), Flow, ELISA
The product encoded by this gene belongs to the actin family of proteins, which are highly conserved proteins that play a role in cell motility, structure and integrity. Alpha, beta and gamma actin isoforms have been identified, with alpha actins being a major constituent of the contractile apparatus, while beta and gamma actins are involved in the regulation of cell motility. This actin is an alpha actin that is found in skeletal muscle. Mutations in this gene cause nemaline myopathy type 3, congenital myopathy with excess of thin myofilaments, congenital myopathy with cores, and congenital myopathy with fiber-type disproportion, diseases that lead to muscle fiber defects.
Specifications
ACTA1 | |
Recombinant Monoclonal | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P68133 | |
ACTA1 | |
A synthesized peptide derived from Human ACTA1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin) | |
4C1 | |
Unconjugated | |
ACTA1 | |
AA959943; ACTA; Acta1; Acta-2; actin alpha 1; actin alpha 1, skeletal muscle; actin, alpha 1, skeletal muscle; actin, alpha skeletal muscle; Actin, alpha skeletal muscle, intermediate form; Acts; Actsk-1; alpha actin 1; alpha-actin-1; ASMA; beta actin; CFTD; CFTD1; CFTDM; cytoplasmic beta-actin; LOW QUALITY PROTEIN: actin, alpha skeletal muscle; MPFD; NEM1; NEM2; NEM3; nemaline myopathy type 3; SHPM; skeletal alpha actin; unnamed protein product | |
Rabbit | |
Affinity Chromatography | |
RUO | |
58 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction