Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACTA1 Recombinant Rabbit Monoclonal Antibody (8B9Q6)
GREENER_CHOICE

Catalog No. PIMA542772
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542772 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA542772 Supplier Invitrogen™ Supplier No. MA542772
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: HeLa, A-431, C6, Mouse lung, Mouse brain, Mouse heart, Rat heart. The target is usually found in the following locations: Cytoplasm, cytoskeleton. Immunogen sequence: ITIGNERFRC PETLFQPSFI GMESAGIHET TYNSIMKCDI DIRKDLYANN VMSGGTTMYP GIADRMQKEI TALAPSTMKI KIIAPPERKY SVWIGGSILA S.

The product encoded by this gene belongs to the actin family of proteins, which are highly conserved proteins that play a role in cell motility, structure and integrity. Alpha, beta and gamma actin isoforms have been identified, with alpha actins being a major constituent of the contractile apparatus, while beta and gamma actins are involved in the regulation of cell motility. This actin is an alpha actin that is found in skeletal muscle. Mutations in this gene cause nemaline myopathy type 3, congenital myopathy with excess of thin myofilaments, congenital myopathy with cores, and congenital myopathy with fiber-type disproportion, diseases that lead to muscle fiber defects.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACTA1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 8B9Q6
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ACTA1
Gene Accession No. P68133, P68134, P68136
Gene Alias AA959943; ACTA; Acta1; Acta-2; actin alpha 1; actin alpha 1, skeletal muscle; actin, alpha 1, skeletal muscle; actin, alpha skeletal muscle; Actin, alpha skeletal muscle, intermediate form; Acts; Actsk-1; alpha actin 1; alpha-actin-1; ASMA; beta actin; CFTD; CFTD1; CFTDM; cytoplasmic beta-actin; LOW QUALITY PROTEIN: actin, alpha skeletal muscle; MPFD; NEM1; NEM2; NEM3; nemaline myopathy type 3; SHPM; skeletal alpha actin; unnamed protein product
Gene Symbols ACTA1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 250-350 of human alpha-Actin-1 (ACTA1) (P68133).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11459, 29437, 58
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.