Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACTA1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112856
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112856 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112856 Supplier Invitrogen™ Supplier No. PA5112856
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ACTA1B Polyclonal Antibody for Western Blot, ELISA

The product encoded by this gene belongs to the actin family of proteins, which are highly conserved proteins that play a role in cell motility, structure and integrity. Alpha, beta and gamma actin isoforms have been identified, with alpha actins being a major constituent of the contractile apparatus, while beta and gamma actins are involved in the regulation of cell motility. This actin is an alpha actin that is found in skeletal muscle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACTA1B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene acta1b
Gene Accession No. 0
Gene Alias acta1b; actc; actc1; actin; actin alpha 1, skeletal muscle b; actin, alpha 1b, skeletal muscle; actin, alpha, cardiac muscle; actin, alpha, cardiac muscle 1; cardiofunk; Cfk; hm:zeh0631; zeh0631
Gene Symbols acta1b
Host Species Rabbit
Immunogen Recombinant Zebrafish Actin protein (1-168AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 407658
Target Species Zebrafish
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.