Learn More
ACTA2/smooth muscle actin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | ACTA2/smooth muscle actin |
---|---|
Concentration | 0.17 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-3992
![]() ![]() |
Proteintech Group Inc
230811AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody is specific to the ACTA2. It selectively stains the vascular smooth muscle but not cardiac or skeletal muscle.
Among the six actin isoforms described in mammals, two are found in virtually all cells (b- and g-cytoplasmic), two are detected in smooth muscle cells (a- and g-smooth muscle) and two are present in striated muscles, one predomitly in skeletal (a-skeletal) and one in cardiac (a-cardiac) muscle cells. These actin isoforms differ slightly in their N-terminus and the sequences of each of them are perfectly conserved in higher vertebrates. Alpha-smooth muscle actin is abundant in vascular and visceral smooth muscle cells. In addition, it has also been shown to appear in stress fibers of fibroblastic cells during pathological situations involving contractile phenomena such as wound healing and fibrocontractive diseases.Specifications
ACTA2/smooth muscle actin | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P62736, P62737, P62738 | |
11475, 59, 81633 | |
ACTA2/smooth muscle actin Fusion Protein Ag19481 | |
Primary | |
-20°C | |
ACTA2 |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
a SMA, AAT6, ACTA2, ACTA2 Smooth muscle Actin, ACTA2 specific, Actin, aortic smooth muscle, ACTSA, ACTVS, Alpha actin 2, Alpha SMA, a-SMA | |
ACTA2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.