Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACTB Mouse anti-Human, Clone: 2A5, Abnova™

Catalog No. 89933513 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-513 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-513 Supplier Abnova Corporation Supplier No. MAB12983
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of human ACTB.

This gene encodes one of six different actin proteins. Actins are highly conserved proteins that are involved in cell motility, structure, and integrity. This actin is a major constituent of the contractile apparatus and one of the two nonmuscle cytoskeletal actins. [provided by RefSeq]

Specifications

Antigen ACTB
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone BA3R
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of human ACTB.
Dilution ELISA Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) Western Blot (1:1000-1:10000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.05% sodium azide).
Gene Alias PS1TP5BP1
Gene Symbols ACTB
Host Species Mouse
Immunogen A synthetic peptide (conjugated with KLH) corresponding to N-terminus region of human ACTB.
Purification Method Affinity purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.