Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACTBL1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988669
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8669 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8669 Supplier Bioss Supplier No. BS8594R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

POTEH belongs to the POTE family and contains 7 ANK repeats.

Specifications

Antigen ACTBL1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene POTEH
Gene Accession No. Q6S545
Gene Alias A26C3; ACTBL1; actin, beta-like 1; ANKRD26-like family C member 3; ANKRD26-like family C, member 3; cancer/testis antigen family 104, member 7; CT104.7; POTE ankyrin domain family member H; POTE ankyrin domain family, member H; POTE22; POTE-22; POTEH; Prostate, ovary, testis-expressed protein on chromosome 22; protein expressed in prostate, ovary, testis, and placenta 22; protein expressed in prostate, ovary, testis, and placenta POTE14 like
Gene Symbols POTEH
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ACTBL1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23784
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.