Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACTBL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572402
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572402 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572402 Supplier Invitrogen™ Supplier No. PA572402
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ACTBL2 Polyclonal Antibody for Western Blot

Actins are highly conserved proteins that are involved in various types of cell motility and are ubiquitously expressed in all eukaryotic cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACTBL2
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Gene ACTBL2
Gene Accession No. Q562R1, Q8BFZ3
Gene Alias 4732495G21Rik; ACT; Actbl2; actin, beta like 2; actin, beta-like 2; actin-like protein; beta-actin-like protein 2; Kappa-actin
Gene Symbols ACTBL2
Host Species Rabbit
Immunogen region 105-131 aa.
Purification Method Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 238880, 345651
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.