Learn More
ACTC1 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1F2B9, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 661251IG150UL
Description
This antibody is specific to the ACTC1. It does not cross-react with other actin isoforms.
ACTC1 are part of actins, which are highly conserved proteins that are involved in various types of cell motility. Polymerization of globular actin (G-actin) leads to a structural filament (F-actin) in the form of a two-stranded helix. Each actin can bind to four others. The protein encoded by this gene belongs to the actin family which is comprised of three main groups of actin isoforms, alpha, beta, and gamma. The alpha actins are found in muscle tissues and are a major constituent of the contractile apparatus. Defects in this gene have been associated with idiopathic dilated cardiomyopathy (IDC) and familial hypertrophic cardiomyopathy (FHC).Specifications
| ACTC1 | |
| Monoclonal | |
| 0.75 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P68032, P68033, P68035 | |
| ACTC1 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Pig, Rat | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1F2B9 | |
| Unconjugated | |
| ACTC1 | |
| ACTC, ACTC1, ACTC1 cardiac muscle 1, ACTC1 specific, Actin cardiac muscle specific, Actin, alpha cardiac muscle 1, actin, alpha, cardiac muscle 1, Alpha cardiac actin, CMD1R, CMH11 | |
| Mouse | |
| Protein G | |
| RUO | |
| 100152267, 11464, 29275, 70 | |
| -20°C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.