Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACTH Antibody (CLIP/1407) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$646.00
Specifications
Antigen | ACTH |
---|---|
Clone | CLIP/1407 |
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry : 0.5-1ug/million cells, ELISA, Immunohistochemistry-Paraffin : 0.5-1ug/ml, Immunofluorescence : 1-2ug/ml, CyTOF-ready |
Applications | Flow Cytometry, ELISA, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP247683A
![]() |
Novus Biologicals
NBP2476830.1MG |
0.1 mg |
Each for $646.00
|
|
|||||
NBP247683B
![]() |
Novus Biologicals
NBP2476830.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
ACTH Monoclonal specifically detects ACTH in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
ACTH | |
1.0 mg/mL | |
Flow Cytometry, ELISA, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF | |
Unconjugated | |
Mouse | |
Neuroscience, Nutrient Sensing in the Brain | |
PBS with No Preservative | |
5443 | |
Synthetic peptide corresponding to aa25-39 of human ACTH (NGAEDESAEAFPLEF) | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
CLIP/1407 | |
Flow Cytometry : 0.5-1ug/million cells, ELISA, Immunohistochemistry-Paraffin : 0.5-1ug/ml, Immunofluorescence : 1-2ug/ml, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
ACTH, adrenocorticotropic hormone, adrenocorticotropin, alpha-melanocyte-stimulating hormone, alpha-MSH, beta-endorphin, beta-LPH, beta-melanocyte-stimulating hormone, beta-MSH, CLIP, corticotropin-like intermediary peptide, corticotropin-lipotropin, gamma-LPH, gamma-MSH, lipotropin beta, lipotropin gamma, LPH, melanotropin alpha, melanotropin beta, melanotropin gamma, met-enkephalin, MSH, NPP, POC, pro-ACTH-endorphin, proopiomelanocortin, pro-opiomelanocortin, proopiomelanocortin preproprotein | |
POMC | |
IgG | |
Protein A or G purified | |
ACTH (same as Corticotropin) is a 39 amino acid active peptide produced by the anterior pituitary. This MAb is specific to CLIP (aa25-39 of ACTH); does not react with Synacthen (aa1-24 of ACTH). POMC (pro-opiomelanocortin or corticotropin-lipotropin) is a 267 amino acid polypeptide hormone precursor that goes through extensive, tissue-specific posttranslational processing by convertases. POMC is cleaved into ten hormone chains named NPP, ACTH, alpha-MSH (Melanocyte Stimulating Hormone), beta-MSH, gamma-MSH, CLIP (corticotropin-like intermediary peptide), Lipotropin-beta, Lipotropin-gamma, beta-endorphin and Met-enkephalin. ACTH is also produced by cells of immune system (T-cells, B-cells, and macrophages) in response to stimuli associated with stress. Anti-ACTH is a useful marker in classification of pituitary tumors and the study of pituitary disease. It reacts with ACTH-producing cells (corticotrophs). It also may react with other tumors (e.g. some small cell carcinomas of the lung) causing paraneoplastic syndromes by secreting ACTH. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title