Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACTH Antibody (rCLIP/1407), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308617G
Description
ACTH Monoclonal antibody specifically detects ACTH in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)Specifications
ACTH | |
Monoclonal | |
DyLight 488 | |
ACTH, adrenocorticotropic hormone, adrenocorticotropin, alpha-melanocyte-stimulating hormone, alpha-MSH, beta-endorphin, beta-LPH, beta-melanocyte-stimulating hormone, beta-MSH, CLIP, corticotropin-like intermediary peptide, corticotropin-lipotropin, gamma-LPH, gamma-MSH, lipotropin beta, lipotropin gamma, LPH, melanotropin alpha, melanotropin beta, melanotropin gamma, met-enkephalin, MSH, NPP, POC, pro-ACTH-endorphin, proopiomelanocortin, pro-opiomelanocortin, proopiomelanocortin preproprotein | |
Synthetic peptide corresponding to (aa 162-176) of human ACTH (NGAEDESAEAFPLEF) (Uniprot: P01189 ) | |
100 μg | |
Neuroscience | |
5443 | |
Store at 4°C in the dark. | |
IgG1 κ |
Immunohistochemistry (Paraffin) | |
rCLIP/1407 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction