Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACTH Mouse anti-Human, Mouse, Rat, Clone: 3C12C7, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | ACTH |
|---|---|
| Clone | 3C12C7 |
| Concentration | 1 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6789
![]()
|
Proteintech Group Inc
663581IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
ATCH (adrenocorticotropic hormone) is a hormone which plays a major role in stimulating the adrenal cortex. It is formed through cleavage of the polypeptide precursor proopiomelanocortin (POMC), which also results in several other cleavage products including MSH, ACTH, and beta endorphin. ATCH is secreted from the anterior pituitary in response to the corticotropin-releasing hormone from the hypothalamus. It stimulates the secretion of glucocorticoids like cortisol, but has little control over the stimulation of mineralocorticoids like aldosterone, which is another major hormone of the adrenal cortex.Specifications
| ACTH | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| ACTH, ACTH Melanotropin alpha, Adrenocorticotropic hormone, Beta LPH Lipotropin gamma, CLIP, CLIP Lipotropin beta, Corticotropin lipotropin, Gamma LPH Melanotropin beta, LPH, MSH, POC, POMC, Pro opiomelanocortin, proopiomelanocortin | |
| Pomc | |
| IgG1 | |
| Protein G | |
| Antibody |
| 3C12C7 | |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| P01189, P01193, P01194 | |
| 18976, 24664, 5443 | |
| ACTH Fusion Protein Ag17311 | |
| Primary | |
| -20°C | |
| POMC |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title