Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACTHR Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981342
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1342 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1342 Supplier Bioss Supplier No. BS11408R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MC2R encodes one member of the five-member G-protein associated melanocortin receptor family. Melanocortins are peptides derived from pro-opiomelanocortin . MC2R is selectively activated by adrenocorticotropic hormone, whereas the other four melanocortin receptors recognize a variety of melanocortin ligands. Mutations in MC2R can result in familial glucocorticoid deficiency.

Specifications

Antigen ACTHR
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MC2R
Gene Accession No. Q01718
Gene Alias ACTH receptor; Acthr; ACTH-R; Adrenocorticotropic hormone receptor; Adrenocorticotropin receptor; corticotropin receptor; MC2 receptor; MC2R; MC2-R; melanocortin 2 receptor; melanocortin 2 receptor (adrenocorticotropic hormone); Melanocortin receptor 2; MGC125798
Gene Symbols MC2R
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MC2 receptor .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4158
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.