Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Actin Alpha 1 Cardiac Muscle Antibody (22D3), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197725AF750
Description
Actin Alpha 1 Cardiac Muscle Monoclonal specifically detects Actin Alpha 1 Cardiac Muscle in Human, Mouse, Rat, Porcine, Bovine, Goat, Mammal, Rabbit, Xenopus, Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting.Specifications
Actin Alpha 1 Cardiac Muscle | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
ACTC1 | |
Clone 22D3 is a mouse monoclonal IgG1 antibody derived by fusion of NS0 mouse myeloma cells with spleen cells from a BALB/c mouse immunized with a peptide comprising the N-terminal nonapeptide of alpha-cardiac actin with an acetylated N-terminus (Ac-DDEETTALVC-COOH) coupled to keyhole limpet hemocyanin through the cysteine residue. | |
0.1 mL | |
Primary | |
Clone 22D3 is an antibody highly specific for a- cardiac actin, and does not cross react with other actin isoforms. | |
Store at 4°C in the dark. |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot | |
22D3 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting | |
ACTC, actin, alpha, cardiac muscle 1, ASD5, CMD1R, CMH11, LVNC4 | |
Mouse | |
Protein A or G purified | |
Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Stem Cell Markers | |
70 | |
Human, Mouse, Rat, Pig, Bovine, Goat, Mammalia, Rabbit, Xenopus species, Zebrafish | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction