Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Actin Alpha 1 Cardiac Muscle Antibody (22D3) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP197725
Description
Actin Alpha 1 Cardiac Muscle Monoclonal specifically detects Actin Alpha 1 Cardiac Muscle in Human, Mouse, Rat, Porcine, Bovine, Goat, Mammal, Rabbit, Xenopus, Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting.Specifications
Actin Alpha 1 Cardiac Muscle | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000-1:5000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen, Immunoblotting 1:1000-1:5000 | |
ACTC1 | |
Clone 22D3 is a mouse monoclonal IgG1 antibody derived by fusion of NS0 mouse myeloma cells with spleen cells from a BALB/c mouse immunized with a peptide comprising the N-terminal nonapeptide of alpha-cardiac actin with an acetylated N-terminus (Ac-DDEETTALVC-COOH) coupled to keyhole limpet hemocyanin through the cysteine residue. | |
0.1 mg | |
Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Stem Cell Markers | |
70 | |
Human, Mouse, Rat, Pig, Bovine, Goat, Mammalia, Rabbit, Xenopus species, Zebrafish | |
IgG1 |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
22D3 | |
Unconjugated | |
ACTC, actin, alpha, cardiac muscle 1, ASD5, CMD1R, CMH11, LVNC4 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Clone 22D3 is an antibody highly specific for a- cardiac actin, and does not cross react with other actin isoforms. | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction