Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

actin alpha-muscle Mouse anti-Chicken, Human, Monkey, Non-human Primate, Porcine, Rabbit, Rat, Zebrafish, Clone: HHF35, MUbio
SDP

Catalog No. 502101011
Encompass
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-210-1011 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-210-1011 Supplier Vector Laboratories Supplier No. MUB0107P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

RCK102 is a Cytokeratin antibody reacting with Cytokeratin 5 and Cytokeratin 8. This monoclonal antibody recognizes virtually all epithelial tissues and carcinomas. The antibody may be stored at 4 C. For prolonged storage prepare appropriate aliquots and store at or below -20 C. Prior to use, an aliquot is thawed slowly in the dark at ambient temperature, spun down again and used to prepare working dilutions by adding sterile phosphate buffered saline (PBS, pH 7.2). Repeated thawing and freezing should be avoided. Working dilutions should be stored at 4 C, not refrozen, and preferably used the same day. If a slight precipitation occurs upon storage, this should be removed by centrifugation. It will not affect the performance or the concentration of the product.

actin alpha-muscle Monoclonal antibody specifically detects actin alpha-muscle in Chicken, Human, Monkey, Non-human Primate, Porcine, Rabbit, Rat, Zebrafish samples. It is validated for ELISA, ELISA, Immunoblot, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen actin alpha-muscle
Applications ELISA, Immunoblot, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone HHF35
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH
Gene ACTG1
Gene Accession No. P63259, P63261, Q5ZMQ2
Gene Alias ACT; ACTA1; ACTB; Actg; ACTG1; actin; actin gamma; actin gamma 1; actin homolog; actin, alpha 1, skeletal muscle; actin, cytoplasmic 2; Actin, cytoplasmic 2, N-terminally processed; actin, cytoplasmic type 5; actin, cytoplasmic, type 5; actin, gamma 1; actin, gamma 1 propeptide; actin, gamma, cytoplasmic 1; Actl; AL023024; Atcb; beta actin; beta-actin; beta-actin FE-3; BRWS2; cytoplasmic; cytoplasmic 2; cytoplasmic ACTG1; cytoplasmic actin; cytoplasmic type 5; cytoskeletal gamma-actin; deafness, autosomal dominant 20; deafness, autosomal dominant 26; DFNA20; DFNA26; E51; epididymis luminal protein 176; gamma 1 propeptide; gamma-actin; HEL-176; RCJMB04_1h13; type 5
Host Species Mouse
Immunogen actin alpha-muscle
Purification Method Protein A/G
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 287876, 397653, 415296, 71
Target Species Chicken, Human, Monkey, Non-human Primate, Pig, Rabbit, Rat, Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.