Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

actin alpha-skeletal Mouse anti-Goat, Human, Porcine, Rabbit, Rat, Clone: 3B3, MUbio
SDP

Catalog No. 502101012
Encompass
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-210-1012 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-210-1012 Supplier Vector Laboratories Supplier No. MUB0108P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

RCK105 reacts exclusively with Cytokeratin 7 which is present in a subgroup of glandular epithelia and their tumors, as well as transitional epithelium and transitional carcinoma. The antibody may be stored at 4 C. For prolonged storage prepare appropriate aliquots and store at or below -20 C. Prior to use, an aliquot is thawed slowly in the dark at ambient temperature, spun down again and used to prepare working dilutions by adding sterile phosphate buffered saline (PBS, pH 7.2). Repeated thawing and freezing should be avoided. Working dilutions should be stored at 4 C, not refrozen, and preferably used the same day. If a slight precipitation occurs upon storage, this should be removed by centrifugation. It will not affect the performance or the concentration of the product.

actin alpha-skeletal Monoclonal antibody specifically detects actin alpha-skeletal in Goat, Human, Porcine, Rabbit, Rat samples. It is validated for ELISA, Immunoblot, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen actin alpha-skeletal
Applications ELISA, Immunoblot, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 3B3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH
Gene ACTA1
Gene Accession No. P68133, P68135, P68136, P68137
Gene Alias AA959943; ACTA; ACTA1; Acta-2; actin alpha 1; actin, alpha 1, skeletal muscle; actin, alpha skeletal muscle; Actin, alpha skeletal muscle, intermediate form; Acts; Actsk-1; Alpha-actin-1; ASMA; CFTD; CFTD1; CFTDM; MPFD; NEM1; NEM2; NEM3; nemaline myopathy type 3; SHPM
Host Species Mouse
Immunogen actin alpha-skeletal
Purification Method Protein A/G
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100009506, 100154254, 102182476, 29437, 58
Target Species Goat, Human, Pig, Rabbit, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.