Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Actin Antibody (3B3), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197723FR
Description
Actin Monoclonal specifically detects Actin in Human, Rat, Porcine, Goat, Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting.Specifications
Actin Alpha 1 Skeletal Muscle | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ACTA1 | |
Clone 3B3 is a mouse monoclonal IgG1 antibody derived by fusion of NS0 mouse myeloma cells with spleen cells from a BALB/c mouse immunized with a peptide comprising the N-terminal nonapeptide of alpha-skeletal actin with an acetylated N-terminus (Ac-DEDETTALVC-COOH) coupled to keyhole limpet hemocyanin through the cysteine residue. | |
0.1 mL | |
Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Stem Cell Markers | |
58 | |
Human, Rat, Pig, Goat, Rabbit | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot | |
3B3 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting | |
ACTA, actin, alpha 1, skeletal muscle, alpha skeletal muscle, alpha skeletal muscle actin, alpha-actin-1, ASMA, CFTD, CFTDM, MPFD, NEM1, NEM2, NEM3 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Clone 3B3 is an antibody highly specific for alpha-skeletal actin, and does not cross react with other actin isoforms. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction