Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Actin Antibody (4C1), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Actin |
---|---|
Clone | 4C1 |
Dilution | Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 |
Applications | Flow Cytometry, ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
Actin Monoclonal antibody specifically detects Actin in Human samples. It is validated for Flow Cytometry, ELISA, ImmunohistochemistrySpecifications
Actin | |
Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
ACTA, actin, alpha 1, skeletal muscle, alpha skeletal muscle, alpha skeletal muscle actin, alpha-actin-1, ASMA, CFTD, CFTDM, MPFD, NEM1, NEM2, NEM3 | |
A synthesized peptide derived from Human Actin [UniProt P68133] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
4C1 | |
Flow Cytometry, ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Angiogenesis, Cancer, Cell Biology, Cellular Markers, Stem Cell Markers | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
58 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title