Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Actin Antibody (mAbGEa) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB013634 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013634 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013634 Supplier Novus Biologicals Supplier No. NBP280561
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Actin Monoclonal specifically detects Actin in Human, Mouse, Rat, Bovine, Chinese Hamster, Drosophila, Fungi, Plant, Protozoa, Rabbit, Sheep, Xenopus, Yeast, Zebrafish samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular), CyTOF-ready.

Specifications

Antigen Actin
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone mAbGEa
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:1000, Simple Western : 1:25, Flow Cytometry : 1:10 - 1:1000, ELISA : 1:100 - 1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence : 1:20 - 1:100, Immunohistochemistry-Paraffin : 1:200, Flow (Intracellular), CyTOF-ready
Formulation PBS
Gene Alias ACTA, actin, alpha 1, skeletal muscle, alpha skeletal muscle, alpha skeletal muscle actin, alpha-actin-1, ASMA, CFTD, CFTDM, MPFD, NEM1, NEM2, NEM3
Gene Symbols ACTA1
Host Species Mouse
Immunogen Purified recombinant Arabidopsis Actin protein [UniProt# P0CJ46]
Purification Method IgM purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, Cellular Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 58
Test Specificity Chinese hamster data from customer review. Sordaria macrospora reactivity reported in scientific literature (PMID: 24720701).
Target Species Human, Mouse, Rat, Bovine, Chinese Hamster, Drosophila, Fungi, Plant, Protozoa, Rabbit, Sheep, Xenopus species, Yeast, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.