Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Actin Antibody (mAbGEa), Novus Biologicals™
SDP

Catalog No. p-4977517 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10074340T 0.025 mL
NB10074340 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10074340T Supplier Novus Biologicals Supplier No. NB10074340SS
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 29 publications

Actin Monoclonal specifically detects Actin in Human, Mouse, Rat, Bovine, Chinese Hamster, Drosophila, Fungi, Plant, Protozoa, Rabbit, Sheep, Xenopus, Yeast, Zebrafish samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).

Specifications

Antigen Actin
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone mAbGEa
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:1000, Simple Western 1:25, Flow Cytometry 1 ug/ml, ELISA 1:100 - 1:2000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunohistochemistry-Paraffin 1:200, Flow (Intracellular) 1 ug/ml
Gene Alias ACTA, actin, alpha 1, skeletal muscle, alpha skeletal muscle, alpha skeletal muscle actin, alpha-actin-1, ASMA, CFTD, CFTDM, MPFD, NEM1, NEM2, NEM3
Gene Symbols ACTA1
Host Species Mouse
Immunogen Purified recombinant Arabidopsis Actin protein [UniProt# P0CJ46]
Purification Method IgM purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 58
Test Specificity Reacts with Actin 1, 2, 3, 4, 7, 8, 11 and 12.
Target Species Human, Mouse, Rat, Bovine, Chinese Hamster, Drosophila, Fungi, Plant, Protozoa, Rabbit, Sheep, Xenopus species, Yeast, Zebrafish
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.