Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Actin Gamma 1 Antibody (2A3), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197720AF405
Description
Actin Gamma 1 Monoclonal specifically detects Actin Gamma 1 in Human, Mouse, Rat, Porcine, Chicken, Rabbit, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting.Specifications
Actin Gamma 1 | |
Monoclonal | |
Alexa Fluor 405 | |
P63261 | |
ACTG1 | |
2A3 is a Mouse monoclonal IgG2b antibody derived by fusion of NS0 Mouse myeloma cells with spleen cells from a BALB/c Mouse immunized with a peptide comprising the N-terminal nonapeptide of gamma-cytoplasmic actin with an acetylated N-terminus (Ac-EEEIAAL | |
0.1 mL | |
Cell Biology, Cellular Markers, Cytoskeleton Markers, Stem Cell Markers | |
71 | |
Human, Mouse, Rat, Pig, Chicken, Mammalia, Rabbit, Zebrafish | |
IgG2b |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot | |
2A3 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting | |
ACT, ACTB, ACTG, actin, cytoplasmic 2, actin, gamma 1, cytoskeletal gamma-actin, deafness, autosomal dominant 20; deafness, autosomal dominant 26, DFNA20, DFNA26, Gamma-actin | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
2A3 is highly specific for gamma-cytoplasmic actin (gamma-actin 1; encoded by ACTG1). It does not cross-react with the gamma-actin isoform expressed in the enteric smooth muscle tissues (gamma-actin 2; encoded by ACTG2) and does not cross react with other | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction