Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Actin Gamma 1 Antibody (NH3), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 6 publications

$268.00 - $560.50

Specifications

Antigen F-Actin
Clone NH3
Concentration 1.0 mg/mL
Dilution Western Blot 1:100-1:500, Flow Cytometry 1:10, ELISA 1:10, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB100D92SS
SDP
View Documents
Novus Biologicals
NB10064792SS
0.05 mg
Each for $268.00
Only null left
Add to Cart
 
NB10064792
SDP
View Documents
Novus Biologicals
NB10064792
0.25 mg
Each for $560.50
Only null left
Add to Cart
 
Description

Description

Actin Gamma 1 Monoclonal specifically detects Actin Gamma 1 in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

F-Actin
1.0 mg/mL
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Unconjugated
Mouse
Human, Mouse, Rat, Rabbit
actin, cytoplasmic 2, DFNA26, ACT, ACTG, actin, gamma 1, BRWS2, DFNA20
ACTG1
IgM
IgM purified
Recognizes human Filamentous actin (F-actin). The binds to the N-terminal region of actin, but not to the extreme N-terminal 40 amino acids. In tissue sections the stains the cytoplasm of macrophages strongly, and gives granular, localized nuclear staining of all cell types.
NH3
Western Blot 1:100-1:500, Flow Cytometry 1:10, ELISA 1:10, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Monoclonal
Purified
RUO
P63261
71
Human monocytes and U937 cell line
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.